missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-tert-butylthiazole, 98%

Catalog No. AAA1411503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1411503 1 g
AAA1411506 5 g
AAA1411514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1411503 Supplier Thermo Scientific Chemicals Supplier No. A1411503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74370-93-7 | C7H12N2S | 156.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74370-93-7
Molecular Formula C7H12N2S
Molecular Weight (g/mol) 156.247
MDL Number MFCD00051644
InChI Key CUWZBHVYLVGOAB-UHFFFAOYSA-N
Synonym 2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
PubChem CID 2734202
IUPAC Name 4-tert-butyl-1,3-thiazol-2-amine
SMILES CC(C)(C)C1=CSC(=N1)N

Specifications

Melting Point 98°C to 103°C
Quantity 1 g
Beilstein 114270
Formula Weight 156.26
Percent Purity 98%
Chemical Name or Material 2-Amino-4-tert-butylthiazole
Hazard Category H302-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only