missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(p-tolyl)thiazole 98.0+%, TCI America™

Catalog No. p-7118011
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26665G 5 g
A266625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26665G Supplier TCI America Supplier No. A26665G
Only null left

Chemical Identifiers

CAS 2103-91-5
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD00170264
InChI Key ARLHWYFAPHJCJT-UHFFFAOYSA-N
Synonym 4-4-methylphenyl-1,3-thiazol-2-amine, 4-p-tolyl thiazol-2-amine, 2-amino-4-p-tolyl thiazole, 4-p-tolyl-thiazol-2-ylamine, 4-p-tolyl thiazol-2-ylamine, 4-4-methylphenyl-2-thiazolamine, 4-p-tolylthiazol-2-amine, 4-4-methylphenyl-1,3-thiazole-2-ylamine, 2-amino-4-4-methylphenyl thiazole, 4-4-methylphenyl-1,3-thiazol-2-ylamine
PubChem CID 244066
IUPAC Name 4-(4-methylphenyl)-1,3-thiazol-2-amine
SMILES CC1=CC=C(C=C1)C2=CSC(=N2)N

Specifications

Melting Point 137°C
Color Yellow
Quantity 5 g
Formula Weight 190.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(p-tolyl)thiazole