missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(p-tolyl)thiazole, 98%

Catalog No. AAL1386306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1386306 5 g
AAL1386303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1386306 Supplier Thermo Scientific Chemicals Supplier No. L1386306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2103-91-5 | C10H10N2S | 190.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2103-91-5
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD00170264
InChI Key ARLHWYFAPHJCJT-UHFFFAOYSA-N
Synonym 4-4-methylphenyl-1,3-thiazol-2-amine, 4-p-tolyl thiazol-2-amine, 2-amino-4-p-tolyl thiazole, 4-p-tolyl-thiazol-2-ylamine, 4-p-tolyl thiazol-2-ylamine, 4-4-methylphenyl-2-thiazolamine, 4-p-tolylthiazol-2-amine, 4-4-methylphenyl-1,3-thiazole-2-ylamine, 2-amino-4-4-methylphenyl thiazole, 4-4-methylphenyl-1,3-thiazol-2-ylamine
PubChem CID 244066
IUPAC Name 4-(4-methylphenyl)-1,3-thiazol-2-amine
SMILES CC1=CC=C(C=C1)C2=CSC(=N2)N

Specifications

Melting Point 134°C to 138°C
Quantity 5 g
Beilstein 143563
Formula Weight 190.27
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(p-tolyl)thiazole
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only