missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylthiophene-3-carboxamide, 98%

Catalog No. AAB2004303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2004303 1 g
AAB2004306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2004303 Supplier Thermo Scientific Chemicals Supplier No. B2004303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4651-97-2 | C6H8N2OS | 156.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4651-97-2
Molecular Formula C6H8N2OS
Molecular Weight (g/mol) 156.203
MDL Number MFCD00052594
InChI Key AXLIJQRGPVJGSO-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-3-thiophenecarboxamide, 3-thiophenecarboxamide, 2-amino-4-methyl, acmc-20a2ps, maybridge1_003931, 2-amino-4-methyl-thiophen-3-carboxamid, 2-amino-4-methyl-3-thiophene carboxamide, 2-azanyl-4-methyl-thiophene-3-carboxamide, 2-amino-4-methyl-thiophene-3-carboxylic acid amide
PubChem CID 293810
IUPAC Name 2-amino-4-methylthiophene-3-carboxamide
SMILES CC1=CSC(=C1C(=O)N)N

Specifications

Melting Point 173°C to 175°C
Quantity 1 g
Formula Weight 156.2
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylthiophene-3-carboxamide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only