missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-methylpyrimidine 97.0+%, TCI America™
$80.16 - $206.90
Chemical Identifiers
| CAS | 108-52-1 |
|---|---|
| Molecular Formula | C5H7N3 |
| Molecular Weight (g/mol) | 109.132 |
| MDL Number | MFCD00006101 |
| InChI Key | GHCFWKFREBNSPC-UHFFFAOYSA-N |
| Synonym | 2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine |
| PubChem CID | 7939 |
| IUPAC Name | 4-methylpyrimidin-2-amine |
| SMILES | CC1=NC(=NC=C1)N |
Chemical Identifiers
| 108-52-1 | |
| 109.132 | |
| GHCFWKFREBNSPC-UHFFFAOYSA-N | |
| 7939 | |
| CC1=NC(=NC=C1)N |
| C5H7N3 | |
| MFCD00006101 | |
| 2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine | |
| 4-methylpyrimidin-2-amine |
Specifications
| 108-52-1 | |
| C5H7N3 | |
| 5 g | |
| GHCFWKFREBNSPC-UHFFFAOYSA-N | |
| 4-methylpyrimidin-2-amine | |
| 7939 | |
| ≥97.0% (T) | |
| 2-Amino-4-methylpyrimidine |
| 159°C | |
| MFCD00006101 | |
| 2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine | |
| CC1=NC(=NC=C1)N | |
| 109.132 | |
| 109.13 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UV6485000
TSCA : No