missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyrimidine, 97%

Catalog No. AAA1608130
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1608130 250 g
AAA1608109 10 g
AAA1608118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1608130 Supplier Thermo Scientific Chemicals Supplier No. A1608130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-52-1 | C5H7N3 | 109.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-52-1
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006101
InChI Key GHCFWKFREBNSPC-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyrimidine, 2-pyrimidinamine, 4-methyl, 6-methyl-2-pyrimidinamine, pyrimidine, 2-amino-4-methyl, 4-methyl-2-pyrimidinamine, 2-amino-4-methyl-pyrimidine, 4-methylpyrimidin-2-ylamine, 4-methylpyrimidine-2-ylamine, 4-methyl-pyrimidin-2-ylamine, 2-amino-6-methylpyrimidine
PubChem CID 7939
IUPAC Name 4-methylpyrimidin-2-amine
SMILES CC1=NC(=NC=C1)N

Specifications

Melting Point 158°C to 161°C
Quantity 250 g
Beilstein 108506
Formula Weight 109.13
Percent Purity 98%
Physical Form Powder or Lumps
Chemical Name or Material 2-Amino-4-methylpyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-591-5
RTECSNumber UV6485000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only