missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%

Catalog No. p-7036127
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54375MD 250 mg
AAH5437503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54375MD Supplier Thermo Scientific Chemicals Supplier No. H54375MD

CAS: 944401-55-2 | C11H18BN3O2 | 235.09 g/mol

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 944401-55-2
Molecular Formula C11H18BN3O2
Molecular Weight (g/mol) 235.09
MDL Number MFCD18072554
InChI Key SYJMHOBGFXCKRG-UHFFFAOYSA-N
PubChem CID 52987906
IUPAC Name 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES CC1=NC(N)=NC=C1B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Synonym 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-ylamine, 2-amino-4-methylpyrimidine-5-boronic acid pinacol ester, 4-methyl-2-aminopyrimidine-5-boronic acid pinacol ester, 2-amino-4-methylpyrimidin-5-yl boronic acid pinacol ester, 4-methyl-5-4, 4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl pyrimidine-2-ylamine, 4-methyl-5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl pyrimidine-2-ylamine
Solubility Information Slightly soluble in water.
Formula Weight 235.09
Percent Purity 96%
Chemical Name or Material 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only