missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzonitrile, 97%, Thermo Scientific™

Chemical Identifiers

CAS 26830-96-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00173706
InChI Key LGNVAEIITHYWCG-UHFFFAOYSA-N
Synonym 2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
PubChem CID 2801276
IUPAC Name 2-amino-4-methylbenzonitrile
SMILES CC1=CC=C(C#N)C(N)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
TL00552EA
View Documents
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

26830-96-6
132.17
LGNVAEIITHYWCG-UHFFFAOYSA-N
2801276
CC1=CC=C(C#N)C(N)=C1
C8H8N2
MFCD00173706
2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
2-amino-4-methylbenzonitrile

Specifications

26830-96-6
97%
MFCD00173706
LGNVAEIITHYWCG-UHFFFAOYSA-N
2-amino-4-methylbenzonitrile
2801276
97%
97.0
C8H8N2
2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
CC1=CC=C(C#N)C(N)=C1
132.17
132.17
2-Amino-4-methylbenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents