missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-methyl-5-nitropyridine 98.0+%, TCI America™
$59.65 - $236.73
Chemical Identifiers
| CAS | 21901-40-6 |
|---|---|
| Molecular Formula | C6H7N3O2 |
| Molecular Weight (g/mol) | 153.141 |
| MDL Number | MFCD00010692 |
| InChI Key | GRBBNZYMXKTQAI-UHFFFAOYSA-N |
| Synonym | 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine |
| PubChem CID | 243165 |
| IUPAC Name | 4-methyl-5-nitropyridin-2-amine |
| SMILES | CC1=CC(=NC=C1[N+](=O)[O-])N |
Chemical Identifiers
| 21901-40-6 | |
| 153.141 | |
| GRBBNZYMXKTQAI-UHFFFAOYSA-N | |
| 243165 | |
| CC1=CC(=NC=C1[N+](=O)[O-])N |
| C6H7N3O2 | |
| MFCD00010692 | |
| 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine | |
| 4-methyl-5-nitropyridin-2-amine |
Specifications
| 21901-40-6 | |
| C6H7N3O2 | |
| 1 g | |
| GRBBNZYMXKTQAI-UHFFFAOYSA-N | |
| 4-methyl-5-nitropyridin-2-amine | |
| 243165 | |
| ≥98.0% (GC,T) | |
| 2-Amino-4-methyl-5-nitropyridine |
| 224°C | |
| MFCD00010692 | |
| 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine | |
| CC1=CC(=NC=C1[N+](=O)[O-])N | |
| 153.141 | |
| 153.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No