missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methyl-5-nitropyridine, 98%, Thermo Scientific Chemicals

$75.74 - $711.42

Chemical Identifiers

CAS 21901-40-6
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00010692
InChI Key GRBBNZYMXKTQAI-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine
PubChem CID 243165
IUPAC Name 4-methyl-5-nitropyridin-2-amine
SMILES CC1=CC(=NC=C1[N+](=O)[O-])N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1972903
View Documents
Thermo Scientific Chemicals
L1972903
1 g
Each for $75.74
Only null left
 
AAL1972906
View Documents
Thermo Scientific Chemicals
L1972906
5 g
Each for $218.90
Only null left
 
AAL1972914
View Documents
Thermo Scientific Chemicals
L1972914
25 g
Each for $711.42
Only null left
 
Description

Description

2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.

Solubility
Soluble in DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Chemical Identifiers

21901-40-6
153.141
GRBBNZYMXKTQAI-UHFFFAOYSA-N
243165
CC1=CC(=NC=C1[N+](=O)[O-])N
C6H7N3O2
MFCD00010692
2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine
4-methyl-5-nitropyridin-2-amine

Specifications

21901-40-6
C6H7N3O2
1 g
2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine
GRBBNZYMXKTQAI-UHFFFAOYSA-N
4-methyl-5-nitropyridin-2-amine
243165
98%
223°C to 225°C
MFCD00010692
137695
Soluble in DMSO.
CC1=CC(=NC=C1[N+](=O)[O-])N
153.141
153.14
2-Amino-4-methyl-5-nitropyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only