missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. p-7117615
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18881G 1 g
A18885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18881G Supplier TCI America Supplier No. A18881G
Only null left

Chemical Identifiers

CAS 6635-86-5
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
InChI Key IKMZGACFMXZAAT-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-3-nitropyridine, 2-amino-3-nitro-4-picoline, 2-pyridinamine, 4-methyl-3-nitro, 2-amino-3-nitro-4-methylpyridine, 4-methyl-3-nitro-pyridin-2-ylamine, 4-methyl-3-nitro-2-pyridylamine, 4-methyl-3-nitro-2-pyridinamine, 4-methyl-3-nitro-2-aminopyridine, 4-methyl-3-nitropyridin-2-amin, 4mrz
PubChem CID 243166
IUPAC Name 4-methyl-3-nitropyridin-2-amine
SMILES CC1=C(C(=NC=C1)N)[N+](=O)[O-]

Specifications

Melting Point 138°C
Color Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methyl-3-nitropyridine