missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxypyridine 98.0+%, TCI America™

Catalog No. A26901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26901G 1 g
A26905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26901G Supplier TCI America Supplier No. A26901G

Chemical Identifiers

CAS 10201-73-7
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD07437849
InChI Key QPHBCOSULYSASF-UHFFFAOYSA-N
Synonym 2-amino-4-methoxypyridine, 4-methoxy-pyridin-2-ylamine, 2-amino-4-methoxylpyridine, 4-methoxy-2-pyridylamine, 2-amino-4-methoxy pyridine, 2-pyridinamine, 4-methoxy, zlchem 190, pubchem5700, pubchem16566, 4-methoxy-2-pyridinamine
PubChem CID 10898715
IUPAC Name 4-methoxypyridin-2-amine
SMILES COC1=CC(N)=NC=C1

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methoxypyridine