missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxypyridine, 95%

CAS: 10201-73-7 | C6H8N2O | 124.14 g/mol

$192.42 - $857.95

Chemical Identifiers

CAS 10201-73-7
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD07437849
InChI Key QPHBCOSULYSASF-UHFFFAOYSA-N
Synonym 2-amino-4-methoxypyridine, 4-methoxy-pyridin-2-ylamine, 2-amino-4-methoxylpyridine, 4-methoxy-2-pyridylamine, 2-amino-4-methoxy pyridine, 2-pyridinamine, 4-methoxy, zlchem 190, pubchem5700, pubchem16566, 4-methoxy-2-pyridinamine
PubChem CID 10898715
IUPAC Name 4-methoxypyridin-2-amine
SMILES COC1=CC(N)=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6612506
View Documents
Thermo Scientific Chemicals
H6612506
5 g
Each for $192.42
Only null left
 
AAH6612514
View Documents
Thermo Scientific Chemicals
H6612514
25 g
Each for $857.95
Only null left
 
Description

Description

2-Amino-4-methoxypyridine is a highly selective inducible nitric oxide synthase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methoxypyridine is a highly selective inducible nitric oxide synthase inhibitors.

Solubility
Soluble in Dimethyl sulfoxide, methanol. Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

10201-73-7
124.14
QPHBCOSULYSASF-UHFFFAOYSA-N
10898715
COC1=CC(N)=NC=C1
C6H8N2O
MFCD07437849
2-amino-4-methoxypyridine, 4-methoxy-pyridin-2-ylamine, 2-amino-4-methoxylpyridine, 4-methoxy-2-pyridylamine, 2-amino-4-methoxy pyridine, 2-pyridinamine, 4-methoxy, zlchem 190, pubchem5700, pubchem16566, 4-methoxy-2-pyridinamine
4-methoxypyridin-2-amine

Specifications

10201-73-7
Yellow
MFCD07437849
UN2811
Air Sensitive
Soluble in Dimethyl sulfoxide,methanol. Slightly soluble in water.
COC1=CC(N)=NC=C1
124.14
124.14
Solid
115°C to 118°C
C6H8N2O
5 g
471837
2-amino-4-methoxypyridine, 4-methoxy-pyridin-2-ylamine, 2-amino-4-methoxylpyridine, 4-methoxy-2-pyridylamine, 2-amino-4-methoxy pyridine, 2-pyridinamine, 4-methoxy, zlchem 190, pubchem5700, pubchem16566, 4-methoxy-2-pyridinamine
QPHBCOSULYSASF-UHFFFAOYSA-N
4-methoxypyridin-2-amine
10898715
95%
2-Amino-4-methoxypyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only