missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(methoxycarbonyl)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride) 98.0+%, TCI America™

Catalog No. p-7118196
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3028200MG 200 mg
A30281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3028200MG Supplier TCI America Supplier No. A3028200MG

Chemical Identifiers

CAS 380430-55-7
Molecular Formula C8H11BClNO4
Molecular Weight (g/mol) 231.439
MDL Number MFCD02258941
InChI Key IDUSDMZTKZZVAW-UHFFFAOYSA-N
Synonym 2-amino-4-methoxycarbonyl phenyl boronic acid hydrochloride, 2-amino-4-methoxycarbonyl phenylboronic acid hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid, hcl, 2-amino-4-methoxycarbonylphenyl boronic acid hydrochloride, 2-amino-4-methoxycarbonylphenylboronic acid hydrochloride, 2-amino-4-methoxycarbonyl boronic acid hydrochloride, 2-borono-5-methoxycarbonyl aniline hydrochloride, 4-methoxycarbonyl-2-aminophenylboronic acid hydrochloride, methyl 3-amino-4-boronobenzoate hydrochloride, 2-amino-4-methoxycarbonyl benzeneboronic acid hydrochloride
PubChem CID 16427083
IUPAC Name (2-amino-4-methoxycarbonylphenyl)boronic acid;hydrochloride
SMILES B(C1=C(C=C(C=C1)C(=O)OC)N)(O)O.Cl

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 200 mg
Formula Weight 231.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(methoxycarbonyl)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride)