missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxybenzothiazole 98.0+%, TCI America™

Catalog No. A24541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24541G 1 g
A24545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24541G Supplier TCI America Supplier No. A24541G

Chemical Identifiers

CAS 5464-79-9
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.225
MDL Number MFCD00005792
InChI Key YEBCRAVYUWNFQT-UHFFFAOYSA-N
Synonym 2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
PubChem CID 21622
IUPAC Name 4-methoxy-1,3-benzothiazol-2-amine
SMILES COC1=C2C(=CC=C1)SC(=N2)N

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 180.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methoxybenzothiazole
RTECSNumber DL2000000
TSCA No