missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxybenzothiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 160350050

 View more versions of this product

Catalog No. AC160350050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Amino-4-methoxybenzothiazole
96.0
Authentic
Glass bottle
MFCD00005792
2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
COC1=C2C(=CC=C1)SC(=N2)N
180.23
180.23
5464-79-9
100.0
96% min. (GC)
C8H8N2OS
5g
YEBCRAVYUWNFQT-UHFFFAOYSA-N
4-methoxy-1,3-benzothiazol-2-amine
21622
97%

Chemical Identifiers

5464-79-9
180.23
YEBCRAVYUWNFQT-UHFFFAOYSA-N
21622
COC1=C2C(=CC=C1)SC(=N2)N
C8H8N2OS
MFCD00005792
2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
4-methoxy-1,3-benzothiazol-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-763-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only