missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxybenzothiazole, 97%

Catalog No. AAL1135814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1135814 25 g
AAL1135803 1 g
AAL1135806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1135814 Supplier Thermo Scientific Chemicals Supplier No. L1135814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5464-79-9 | C8H8N2OS | 180.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5464-79-9
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.225
MDL Number MFCD00005792
InChI Key YEBCRAVYUWNFQT-UHFFFAOYSA-N
Synonym 2-amino-4-methoxybenzothiazole, 4-methoxy-2-aminobenzothiazole, 2-benzothiazolamine, 4-methoxy, 4-methoxybenzo d thiazol-2-amine, 2-amino-4-methoxy-benzothiazole, benzothiazole, 2-amino-4-methoxy, 4-methoxy-benzothiazol-2-ylamine, 4-methoxybenzothiazol-2-ylamine, 4-methoxybenzothiazole-2-ylamine, 2-benzothiazolamine, 4-methoxy-9ci
PubChem CID 21622
IUPAC Name 4-methoxy-1,3-benzothiazol-2-amine
SMILES COC1=C2C(=CC=C1)SC(=N2)N

Specifications

Melting Point 153°C to 159°C
Quantity 25 g
Beilstein 141363
Formula Weight 180.23
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methoxybenzothiazole
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 226-763-1
RTECSNumber DL2000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only