missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-imino-2-thiazoline hydrochloride, 99%

Catalog No. AAL0584406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0584406 5 g
AAL0584414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0584406 Supplier Thermo Scientific Chemicals Supplier No. L0584406

CAS: 36518-76-0 | C3H6ClN3S | 151.612 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36518-76-0
Molecular Formula C3H6ClN3S
Molecular Weight (g/mol) 151.612
MDL Number MFCD00012721
InChI Key ANDLBJXZBLTCDP-UHFFFAOYSA-N
Synonym 2-amino-4-imino-2-thiazoline hydrochloride, 2,4-diiminothiazolidine hydrochloride, 2-imino-2,5-dihydrothiazol-4-amine hydrochloride, 2-imino-5h-1,3-thiazol-4-amine hydrochloride, 4-imino-5h-1,3-thiazol-2-amine hydrochloride, acmc-1aeug, 4-imino-1,3-thiazoline-2-ylamine, chloride, 4-imino-4,5-dihydrothiazol-2-amine hydrochloride
PubChem CID 2723857
IUPAC Name 2-imino-5H-1,3-thiazol-4-amine;hydrochloride
SMILES C1C(=NC(=N)S1)N.Cl

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 5109728
Formula Weight 151.62
Percent Purity 99%
Chemical Name or Material 2-Amino-4-imino-2-thiazoline hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 253-083-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only