missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine, 97%

Catalog No. AAA1853303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1853303 1 g
AAA1853306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1853303 Supplier Thermo Scientific Chemicals Supplier No. A1853303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1513-69-5 | C5H4F3N3O | 179.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1513-69-5
Molecular Formula C5H4F3N3O
Molecular Weight (g/mol) 179.102
MDL Number MFCD00068093
InChI Key ZEPSVMLZBXDPGU-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxy-6-trifluoromethyl pyrimidine, 2-amino-6-trifluoromethyl pyrimidin-4-ol, 2-amino-6-trifluoromethyl-pyrimidin-4-ol, 2-amino-6-trifluoromethyl-1h-pyrimidin-4-one, 2-amino-4-hydroxy-6-trifluoromethylpyrimidine, 2-amino-4-hydroxy-6-trifluoromethyl-pyrimidine, 4 1h-pyrimidinone, 2-amino-6-trifluoromethyl, pubchem6976, acmc-1bvtd, maybridge1_007193
PubChem CID 233891
IUPAC Name 2-amino-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES C1=C(NC(=NC1=O)N)C(F)(F)F

Specifications

Melting Point 288°C to 292°C
Quantity 1 g
Beilstein 3608100
Formula Weight 179.1
Percent Purity 97%
Chemical Name or Material 2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only