missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxy-6-methylpyrimidine 98.0+%, TCI America™

Catalog No. p-7117308
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A120425G 25 g
A1204250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A120425G Supplier TCI America Supplier No. A120425G
Only null left

Chemical Identifiers

CAS 3977-29-5
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.131
MDL Number MFCD00006095
InChI Key KWXIPEYKZKIAKR-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
PubChem CID 1532
ChEBI CHEBI:58959
IUPAC Name 2-amino-6-methyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 125.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-hydroxy-6-methylpyrimidine