missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxy-6-methylpyrimidine, 99%

Catalog No. AC103831000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103831000 100 g Plastic bottle
AC103830250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103831000 Supplier Thermo Scientific Chemicals Supplier No. 103831000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3977-29-5 | C5H7N3O | 125.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3977-29-5
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.13
MDL Number MFCD00006095
InChI Key KWXIPEYKZKIAKR-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
PubChem CID 1532
ChEBI CHEBI:58959
IUPAC Name 2-amino-6-methyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 24, 343
Solubility Information Solubility in water: insoluble.
Formula Weight 125.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Amino-4-hydroxy-6-methylpyrimidine, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-612-1
RTECSNumber UW7362250

RUO – Research Use Only