missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxy-6-methylpyrimidine, 98%

Catalog No. p-7027669
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2460418 50 g
AAB2460430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2460418 Supplier Thermo Scientific Chemicals Supplier No. B2460418
Only null left

CAS: 3977-29-5 | C5H7N3O | 125.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3977-29-5
Molecular Formula C5H7N3O
Molecular Weight (g/mol) 125.131
MDL Number MFCD00006095
InChI Key KWXIPEYKZKIAKR-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxy-6-methylpyrimidine, 2-amino-6-methylpyrimidin-4-ol, 6-methylisocytosine, mecytosine, 2-amino-6-methylpyrimidin-4 3h-one, superacil, superacyl, 4-methylisocytosine, 2-amino-6-hydroxy-4-methylpyrimidine, 2-amino-6-methyl-4-pyrimidinol
PubChem CID 1532
ChEBI CHEBI:58959
IUPAC Name 2-amino-6-methyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)N

Specifications

Melting Point ∼305°C (decomposition)
Quantity 50 g
Beilstein 3531
Formula Weight 125.13
Percent Purity 98%
Chemical Name or Material 2-Amino-4-hydroxy-6-methylpyrimidine
EINECSNumber 223-612-1
RTECSNumber UW7362250
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only