Learn More
2-Amino-4-ethylpyridine, 97%
CAS: 33252-32-3 | C7H11N2 | 123.18 g/mol
Supplier: Thermo Scientific Chemicals A1875409
Description
2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-ethylpyridine is used as a precursor to prepare 1-ethyl-3-[5-(7-ethyl-imidazo[1,2-a]pyridin-2-yl)-2-hydroxy-phenyl]-urea. It is also used as an intermediate in organic synthesis. It is considered as an analogue of pirlimycin, which acts as an antibiotic.
Solubility
Soluble in water, hexane, ethyl acetate and methyl tert-butyl ether.
Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications
| 2-Amino-4-ethylpyridine | |
| 65°C to 69°C | |
| MFCD00129025 | |
| UN2811 | |
| Soluble in water,hexane,ethyl acetate and methyl tert-butyl ether. | |
| CCC1=CC=[NH+]C(N)=C1 | |
| 123.18 | |
| 122.17 |
| 33252-32-3 | |
| C7H11N2 | |
| 10 g | |
| 2-amino-4-ethylpyridine, 4-ethyl-pyridin-2-ylamine, 4-ethyl-2-pyridinamine, 2-pyridinamine, 4-ethyl, chembl62077, pubchem7108, 4-ethyl-2-pyridylamine, 4-ethyl-2-aminopyridine, acmc-20ao52 | |
| SJWHILBZPGQBJE-UHFFFAOYSA-O | |
| 4-ethylpyridin-2-amine | |
| 118425 | |
| 97% |
Chemical Identifiers
| 33252-32-3 | |
| 123.18 | |
| SJWHILBZPGQBJE-UHFFFAOYSA-O | |
| 118425 | |
| CCC1=CC=[NH+]C(N)=C1 |
| C7H11N2 | |
| MFCD00129025 | |
| 2-amino-4-ethylpyridine, 4-ethyl-pyridin-2-ylamine, 4-ethyl-2-pyridinamine, 2-pyridinamine, 4-ethyl, chembl62077, pubchem7108, 4-ethyl-2-pyridylamine, 4-ethyl-2-aminopyridine, acmc-20ao52 | |
| 4-ethylpyridin-2-amine |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 251-430-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only