missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methylpyrimidine 98.0+%, TCI America™

Catalog No. A10885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10885G 5 g
A108825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10885G Supplier TCI America Supplier No. A10885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5600-21-5
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.574
MDL Number MFCD00006091
InChI Key NPTGVVKPLWFPPX-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methylpyrimidine, am inhibitor, am pesticide, prepn. am, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, 2-pyrimidinamine, 4-chloro-6-methyl, am nitrification inhibitor, pyrimidine, 2-amino-4-chloro-6-methyl, 2-amino-6-chloro-4-methylpyrimidine
PubChem CID 21810
ChEBI CHEBI:58960
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
SMILES CC1=CC(=NC(=N1)N)Cl

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 5 g
Formula Weight 143.57
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine
EINECSNumber (5)-0928
RTECSNumber UV6329600
TSCA No