missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-6-methylpyrimidine, 98%

Catalog No. AAL0689606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0689606 5 g
AAL0689614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0689606 Supplier Thermo Scientific Chemicals Supplier No. L0689606

CAS: 5600-21-5 | C5H6ClN3 | 143.574 g/mol

2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloro-6-methylpyrimidine is a nitrification inhibitor. It was used to study the influence of chlorine substitution in pyrimidine ring on proton donor ability of amino group in 2-aminopyrimidine.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5600-21-5
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.574
MDL Number MFCD00006091
InChI Key NPTGVVKPLWFPPX-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methylpyrimidine, am inhibitor, am pesticide, prepn. am, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, 2-pyrimidinamine, 4-chloro-6-methyl, am nitrification inhibitor, pyrimidine, 2-amino-4-chloro-6-methyl, 2-amino-6-chloro-4-methylpyrimidine
PubChem CID 21810
ChEBI CHEBI:58960
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
SMILES CC1=CC(=NC(=N1)N)Cl

Specifications

Melting Point 181°C to 186°C
Quantity 5 g
Beilstein 114297
Solubility Information Insoluble in water.
Formula Weight 143.58
Percent Purity 98%
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-018-3
RTECSNumber UV6329600
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only