missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-3,5,6-trifluoropyridine, 98%

Catalog No. AAH3415503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3415503 1 g
AAH3415506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3415503 Supplier Thermo Scientific Chemicals Supplier No. H3415503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63489-56-5 | C5H2ClF3N2 | 182.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63489-56-5
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
MDL Number MFCD08436088
InChI Key WZRMLGNDGPCZFG-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-3,5,6-trifluoropyridine, 4-chloro-3,5,6-trifluoro-2-pyridinamine, 4-chloro-3,5,6-trifluoro-2-pyridylamine, 3,5,6-trifluoro-4-chloropyridine-2-amine, 4-chloranyl-3,5,6-tris fluoranyl pyridin-2-amine
PubChem CID 12508075
IUPAC Name 4-chloro-3,5,6-trifluoropyridin-2-amine
SMILES C1(=C(C(=C(N=C1N)F)F)Cl)F

Specifications

Melting Point 119°C to 123°C
Quantity 1 g
Formula Weight 182.54
Percent Purity 98%
Chemical Name or Material 2-Amino-4-chloro-3,5,6-trifluoropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only