missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dimethylpyrimidine 98.0+%, TCI America™

Catalog No. A108325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A108325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A108325G Supplier TCI America Supplier No. A108325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 767-15-7
Melting Point 153°C
Color White-Yellow
Molecular Formula C6H9N3
Quantity 25 g
MDL Number MFCD00006102
Synonym 2-amino-4,6-dimethylpyrimidine, acetylacetoneguanidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, 4,6-dimethyl-2-aminopyrimidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m, 2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine
InChI Key IDQNBVFPZMCDDN-UHFFFAOYSA-N
SMILES CC1=CC(C)=NC(N)=N1
IUPAC Name 4,6-dimethylpyrimidin-2-amine
Molecular Weight (g/mol) 123.16
PubChem CID 13021
Formula Weight 123.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-dimethylpyrimidine
Show More Show Less