missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dimethylpyrimidine, 98%

Catalog No. AAA1367422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1367422 100 g
AAA1367414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1367422 Supplier Thermo Scientific Chemicals Supplier No. A1367422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 767-15-7 | C6H9N3 | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 767-15-7
Molecular Formula C6H9N3
Molecular Weight (g/mol) 123.16
MDL Number MFCD00006102
InChI Key IDQNBVFPZMCDDN-UHFFFAOYSA-N
Synonym 2-amino-4,6-dimethylpyrimidine, acetylacetoneguanidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, 4,6-dimethyl-2-aminopyrimidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m, 2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine
PubChem CID 13021
IUPAC Name 4,6-dimethylpyrimidin-2-amine
SMILES CC1=CC(C)=NC(N)=N1

Specifications

Melting Point 150°C to 155°C
Quantity 100 g
Beilstein 110735
Formula Weight 123.16
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Amino-4,6-dimethylpyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-181-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only