missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dihydroxypyrimidine, 98%

Catalog No. p-54902
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103510250 25 g Glass bottle
AC103511000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103510250 Supplier Thermo Scientific Chemicals Supplier No. 103510250
Only null left

CAS: 56-09-7 | C4H5N3O2 | 127.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-09-7
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD00006094
InChI Key AUFJTVGCSJNQIF-UHFFFAOYSA-N
Synonym 2-amino-4,6-dihydroxypyrimidine, 2-aminopyrimidine-4,6-diol, 4 1h-pyrimidinone, 2-amino-6-hydroxy, 2-amino-6-hydroxypyrimidin-4 3h-one, 2-amino-4,6-dioxypyrimidine, 2-amino-4,6-pyrimidinedione, 4,6-dihydroxy-2-aminopyrimidine, 2-amino-4,6-pyrimidinediol, 2-amino-6-hydroxy-1h-pyrimidin-4-one, 2-amino-6-hydroxy-4 1h-pyrimidinone
PubChem CID 66131
IUPAC Name 2-amino-4-hydroxy-1H-pyrimidin-6-one
SMILES NC1=NC(O)=CC(=O)N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Pink to White or Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 24, 468
Formula Weight 127.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4, 6-dihydroxypyrimidine, 98%
Show More Show Less
EINECSNumber 200-256-5
RTECSNumber UW7361000
TSCA TSCA

RUO – Research Use Only