missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-difluorobenzothiazole, 97%

Catalog No. p-7052724
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5550603 1 g
AAH5550606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5550603 Supplier Thermo Scientific Chemicals Supplier No. H5550603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119256-40-5 | C7H4F2N2S | 186.18 g/mol

2-Amino-4,6-difluorobenzothiazole is used as drugs, food, cosmetics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4,6-difluorobenzothiazole is used as drugs, food, cosmetics

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 119256-40-5
Molecular Formula C7H4F2N2S
Molecular Weight (g/mol) 186.18
MDL Number MFCD00276099
InChI Key DDKKXSCVPKDRRS-UHFFFAOYSA-N
Synonym 2-amino-4,6-difluorobenzothiazole, 4,6-difluorobenzothiazol-2-ylamine, 4,6-difluorobenzo d thiazol-2-amine, 4,6-difluoro-benzothiazol-2-ylamine, 2-benzothiazolamine, 4,6-difluoro, 2-amino-4,6-difluoro-1,3-benzothiazol, 2-amino-4,6-difluoro-1,3-benzothiazole, 4,6-difluorobenzothiazole-2-ylamine, pubchem21756, maybridge1_001141
PubChem CID 737406
IUPAC Name 4,6-difluoro-1,3-benzothiazol-2-amine
SMILES C1=C(C=C2C(=C1F)N=C(S2)N)F

Specifications

Melting Point 216°C to 220°C
Color Brown to White
Quantity 1 g
Formula Weight 186.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-4,6-difluorobenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only