missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloropyrimidine 95.0+%, TCI America™

Catalog No. A145625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A145625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A145625G Supplier TCI America Supplier No. A145625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56-05-3
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.989
MDL Number MFCD00006090
InChI Key JPZOAVGMSDSWSW-UHFFFAOYSA-N
Synonym 2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351
PubChem CID 65522
IUPAC Name 4,6-dichloropyrimidin-2-amine
SMILES C1=C(N=C(N=C1Cl)N)Cl

Specifications

Quantity 25 g
Formula Weight 163.99
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-dichloropyrimidine
RTECSNumber UV6260485
TSCA Yes