missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloropyrimidine, 98%

CAS: 56-05-3 | C4H3Cl2N3 | 163.989 g/mol

Supplier:  Thermo Scientific Chemicals A1133014

 View more versions of this product

Catalog No. AAA1133014


Only null left
Explore available promotions

Description

Description

Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.

Solubility
Insoluble in water, soluble in acetone, two monomer (VCM) and thermal toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Amino-4,6-dichloropyrimidine
218°C to 222°C
MFCD00006090
118454
Insoluble in water,soluble in acetone,two monomer (VCM) and thermal toluene.
C1=C(N=C(N=C1Cl)N)Cl
163.989
164
56-05-3
C4H3Cl2N3
25 g
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351
JPZOAVGMSDSWSW-UHFFFAOYSA-N
4,6-dichloropyrimidin-2-amine
65522
98%

Chemical Identifiers

56-05-3
163.989
JPZOAVGMSDSWSW-UHFFFAOYSA-N
65522
C1=C(N=C(N=C1Cl)N)Cl
C4H3Cl2N3
MFCD00006090
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351
4,6-dichloropyrimidin-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-253-9

RTECSNumber : UV6260485

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only