missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloropyrimidine, 95%, tech.

Catalog No. AC103490500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC103490500 50 g
AC103490100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC103490500 Supplier Thermo Scientific Chemicals Supplier No. 103490500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-05-3 | C4H3Cl2N3 | 163.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-05-3
Molecular Formula C4H3Cl2N3
Molecular Weight (g/mol) 163.99
MDL Number MFCD00006090
InChI Key JPZOAVGMSDSWSW-UHFFFAOYSA-N
Synonym 2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351
PubChem CID 65522
IUPAC Name 4,6-dichloropyrimidin-2-amine
SMILES C1=C(N=C(N=C1Cl)N)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 219°C to 226°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 50 g
Beilstein 24, 81
Formula Weight 163.99
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2-Amino-4, 6-dichloropyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 200-253-9

RUO – Research Use Only