missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde, 96%, Thermo Scientific Chemicals

Catalog No. p-7030284
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2784406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2784406 Supplier Thermo Scientific Chemicals Supplier No. H2784406

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5604-46-6
Molecular Formula C5H3Cl2N3O
Molecular Weight (g/mol) 192.00
MDL Number MFCD03001242
InChI Key GOJUJUVQIVIZAV-UHFFFAOYSA-N
Synonym 2-amino-4,6-dichloro-5-formylpyrimidine, 2-amino-4,6-dichloro-5-pyrimidinecarbaldehyde, 2-amino-4,6-dichloro-5-formyl-pyrimidine, 2-amino-4.6-dichloropyrimidine-5-carbaldehyde, 2-amino-4,6-dichloro-pyrimidine-5-carbaldehyde, 2-amino-4,6-dichloropyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-amino-4,6-dichloro, 2-amino, pubchem6979, acmc-209lqn
PubChem CID 265546
IUPAC Name 2-amino-4,6-dichloropyrimidine-5-carbaldehyde
SMILES NC1=NC(Cl)=C(C=O)C(Cl)=N1

Specifications

Melting Point ∼220°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 192
Percent Purity 96%
Chemical Name or Material 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only