missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloro-5-formamidopyrimidine 98.0+%, TCI America™

Catalog No. A21465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21465G 5 g
A214625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21465G Supplier TCI America Supplier No. A21465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 171887-03-9
Molecular Formula C5H4Cl2N4O
Molecular Weight (g/mol) 207.01
MDL Number MFCD04112936
InChI Key XYWHZUCZNRMJGO-UHFFFAOYSA-N
Synonym N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide
PubChem CID 10198177
IUPAC Name N-(2-amino-4,6-dichloropyrimidin-5-yl)formamide
SMILES NC1=NC(Cl)=C(NC=O)C(Cl)=N1

Specifications

Quantity 5 g
Formula Weight 207.01
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-dichloro-5-formamidopyrimidine