missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,6-dichloro-5-formamidopyrimidine 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 207.01 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-4,6-dichloro-5-formamidopyrimidine |
Safety and Handling
| TSCA | No |