missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-4',5'-dimethoxyacetophenone, 98%

Catalog No. AAL1060506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1060506 5 g
AAL1060514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1060506 Supplier Thermo Scientific Chemicals Supplier No. L1060506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4101-30-8 | C10H13NO3 | 195.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4101-30-8
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00016646
InChI Key KGKWXEGYKGTMAK-UHFFFAOYSA-N
Synonym 1-2-amino-4,5-dimethoxyphenyl ethanone, 2'-amino-4',5'-dimethoxyacetophenone, 2-amino-4,5-dimethoxyacetophenone, 1-2-amino-4,5-dimethoxyphenyl ethan-1-one, 1-2-amino-4,5-dimethoxyhenyl ethanone, ethanone, 1-2-amino-4,5-dimethoxyphenyl, 1-acetyl-2-amino-4,5-dimethoxybenzene, pubchem4007, acmc-1arib, 2-acetyl-4,5-dimethoxyaniline
PubChem CID 602085
IUPAC Name 1-(2-amino-4,5-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1N)OC)OC

Specifications

Melting Point 104°C to 108°C
Quantity 5 g
Beilstein 391380
Formula Weight 195.22
Percent Purity 98%
Chemical Name or Material 2'-Amino-4',5'-dimethoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only