missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,5-dicyano-1H-imidazole 97.0+%, TCI America™
Supplier: TCI America A129225G
Specifications
| 2-Amino-4,5-dicyano-1H-imidazole | |
| C5H3N5 | |
| 25 g | |
| MLOXIXGLIZLPDP-UHFFFAOYSA-N | |
| 2-amino-1H-imidazole-4,5-dicarbonitrile | |
| 170446 | |
| ≥97.0% (T) |
| 40953-34-2 | |
| MFCD00129982 | |
| 2-Amino-4,5-imidazoledicarbonitrile | |
| NC1=NC(C#N)=C(N1)C#N | |
| 133.11 | |
| 133.11 | |
| Crystalline Powder |
Chemical Identifiers
| 40953-34-2 | |
| 133.11 | |
| MLOXIXGLIZLPDP-UHFFFAOYSA-N | |
| 170446 | |
| NC1=NC(C#N)=C(N1)C#N |
| C5H3N5 | |
| MFCD00129982 | |
| 2-Amino-4,5-imidazoledicarbonitrile | |
| 2-amino-1H-imidazole-4,5-dicarbonitrile |
Safety and Handling
EINECSNumber : (5)-5325
TSCA : Yes