missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%

Catalog No. p-7031459
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3254409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3254409 Supplier Thermo Scientific Chemicals Supplier No. H3254409

CAS: 4651-91-6 | C9H10N2S | 178.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4651-91-6
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.253
MDL Number MFCD00128278
InChI Key ADHVMGAFAKSNOM-UHFFFAOYSA-N
Synonym 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitrile, 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitryle, cambridge id 5130181, aminotetrahydrobenzothiophenecarbonitrile
PubChem CID 78382
IUPAC Name 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
SMILES C1CCC2=C(C1)C(=C(S2)N)C#N

Specifications

Melting Point 141°C to 144°C
Quantity 10 g
Formula Weight 178.26
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-085-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only