missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(4-chlorophenyl)thiazole 98.0+%, TCI America™

Catalog No. p-7118103
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A28411G 1 g
A28415G 5 g
2 options

Catalog No. A28411G

Supplier: TCI America A28411G

Only null left

Chemical Identifiers

CAS 2103-99-3
Molecular Formula C9H7ClN2S
Molecular Weight (g/mol) 210.68
MDL Number MFCD00047058
InChI Key DWGWNNCHJPKZNC-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl thiazol-2-amine, 4-4-chlorophenyl-1,3-thiazol-2-amine, 2-amino-4-4-chlorophenyl thiazole, 2-thiazolamine, 4-4-chlorophenyl, 4-4-chloro-phenyl-thiazol-2-ylamine, 4-4-chlorophenyl-2-thiazolamine, 2-amino 4 p-chloro phenyl thiazole, 4-4-chlorophenyl-1,3-thiazole-2-ylamine, p-chloro phenyl-4 amino-2 thiazole french, pubchem9961
PubChem CID 73241
IUPAC Name 4-(4-chlorophenyl)-1,3-thiazol-2-amine
SMILES NC1=NC(=CS1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 210.68
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(4-chlorophenyl)thiazole
RTECSNumber XJ1026732
TSCA No