missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(4-chlorophenyl)-5-methylthiazole 98.0+%, TCI America™

Catalog No. A30661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30661G 1 g
A30665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30661G Supplier TCI America Supplier No. A30661G
Only null left

Chemical Identifiers

CAS 82632-77-7
Molecular Formula C10H9ClN2S
Molecular Weight (g/mol) 224.706
MDL Number MFCD00731832
InChI Key IEJGVSUOFMBPJU-UHFFFAOYSA-N
PubChem CID 667574
IUPAC Name 4-(4-chlorophenyl)-5-methyl-1,3-thiazol-2-amine
SMILES CC1=C(N=C(S1)N)C2=CC=C(C=C2)Cl

Specifications

Melting Point 146°C
Color Yellow
Quantity 1 g
Formula Weight 224.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(4-chlorophenyl)-5-methylthiazole