missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,4,4-trifluorobutyric Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117391
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1367100MG 100 mg
A13671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1367100MG Supplier TCI America Supplier No. A1367100MG
Only null left

Chemical Identifiers

CAS 15959-93-0
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.092
MDL Number MFCD00671488
InChI Key AQPCXCOPDSEKQT-UHFFFAOYSA-N
Synonym 2-amino-4,4,4-trifluorobutyric acid, butanoic acid, 2-amino-4,4,4-trifluoro, s-2-amino-4,4,4-trifluorobutyric acid, 3-trifluoromethylalanine, 2-amino-4,4,4-trifluoro-butyric acid, butanoicacid,2-amino-4,4,4-trifluoro, butanoic acid,2-amino-4,4,4-trifluoro, 2-amino-4,4,4-trifluoro-n-butyric acid hydrochlori
PubChem CID 353083
IUPAC Name 2-amino-4,4,4-trifluorobutanoic acid
SMILES C(C(C(=O)O)N)C(F)(F)F

Specifications

Quantity 100 mg
Formula Weight 193.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,4,4-trifluorobutyric Acid Hydrochloride