missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitropyridine 98.0+%, TCI America™

Catalog No. A08385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A08385G Supplier TCI America Supplier No. A08385G

Chemical Identifiers

CAS 4214-75-9
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006314
InChI Key BPYHGTCRXDWOIQ-UHFFFAOYSA-N
Synonym 2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
PubChem CID 77887
IUPAC Name 3-nitropyridin-2-amine
SMILES NC1=NC=CC=C1[N+]([O-])=O

Specifications

Melting Point 165°C
Color Yellow
Quantity 5 g
Formula Weight 139.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-nitropyridine