missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitropyridine, 99%, Thermo Scientific™

$91.98 - $312.10

Chemical Identifiers

CAS 4214-75-9
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006314
InChI Key BPYHGTCRXDWOIQ-UHFFFAOYSA-N
Synonym 2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
PubChem CID 77887
IUPAC Name 3-nitropyridin-2-amine
SMILES NC1=NC=CC=C1[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC151120050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC151120250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4214-75-9
139.11
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
77887
NC1=NC=CC=C1[N+]([O-])=O
C5H5N3O2
MFCD00006314
2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
3-nitropyridin-2-amine

Specifications

4214-75-9
100.0
Authentic
C5H5N3O2
MFCD00006314
2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
3-nitropyridin-2-amine
77887
99%
98.5
167°C
98.5% min. (GC)
NH2(C5H3N)NO2
22, I, 631
Solubility in water: 3g/L (20 c)
NC1=NC=CC=C1[N+]([O-])=O
139.11
139.11
2-Amino-3-nitropyridine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 224-144-0

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only