missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitropyridine, 99%

Catalog No. AAA1436822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1436822 100 g
AAA1436806 5 g
AAA1436814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1436822 Supplier Thermo Scientific Chemicals Supplier No. A1436822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4214-75-9 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4214-75-9
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006314
InChI Key BPYHGTCRXDWOIQ-UHFFFAOYSA-N
Synonym 2-amino-3-nitropyridine, 3-nitro-2-aminopyridine, 2-pyridinamine, 3-nitro, 3-nitropyridin-2-ylamine, 3-nitro-pyridin-2-ylamine, pyridine, 2-amino-3-nitro, 2-amino3-nitropyridine, 3-nitro-2-pyridinamine, 2-amino-3-nitro pyridine, 3-nitro-2-pyridylamine
PubChem CID 77887
IUPAC Name 3-nitropyridin-2-amine
SMILES NC1=NC=CC=C1[N+]([O-])=O

Specifications

Melting Point 165°C to 167°C
Quantity 100 g
Beilstein 124468
Formula Weight 139.11
Percent Purity 99%
Chemical Name or Material 2-Amino-3-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-144-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only