missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-nitrophenol 98.0+%, TCI America™
$57.75 - $154.02
Chemical Identifiers
| CAS | 603-85-0 |
|---|---|
| Molecular Formula | C6H6N2O3 |
| Molecular Weight (g/mol) | 154.13 |
| MDL Number | MFCD00010875 |
| InChI Key | KUCWUAFNGCMZDB-UHFFFAOYSA-N |
| Synonym | phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol |
| PubChem CID | 4115495 |
| IUPAC Name | 2-amino-3-nitrophenol |
| SMILES | NC1=C(O)C=CC=C1[N+]([O-])=O |
Chemical Identifiers
| 603-85-0 | |
| 154.13 | |
| KUCWUAFNGCMZDB-UHFFFAOYSA-N | |
| 4115495 | |
| NC1=C(O)C=CC=C1[N+]([O-])=O |
| C6H6N2O3 | |
| MFCD00010875 | |
| phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol | |
| 2-amino-3-nitrophenol |
Specifications
| 603-85-0 | |
| C6H6N2O3 | |
| 5 g | |
| KUCWUAFNGCMZDB-UHFFFAOYSA-N | |
| 2-amino-3-nitrophenol | |
| 4115495 | |
| ≥98.0% (HPLC,T) | |
| 2-Amino-3-nitrophenol |
| 209°C | |
| MFCD00010875 | |
| phenol, 2-amino-3-nitro, 2-hydroxy-6-nitroaniline, ccris 2544, unii-a7f97zpu2w, 3-nitro-2-aminophenol, 2-amino-3-nitro-phenol, a7f97zpu2w, 2-amino-3-hydroxynitrobenzene, 1-hydroxy-2-amino-3-nitrobenzene, 2-amino-nitrophenol | |
| NC1=C(O)C=CC=C1[N+]([O-])=O | |
| 154.13 | |
| 154.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0788
TSCA : No