missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylpyridine 98.0+%, TCI America™

Catalog No. p-7116877
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A040025G 25 g
A0400500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A040025G Supplier TCI America Supplier No. A040025G

Chemical Identifiers

CAS 1603-40-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006320
InChI Key RGDQRXPEZUNWHX-UHFFFAOYSA-N
Synonym 2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
PubChem CID 15347
IUPAC Name 3-methylpyridin-2-amine
SMILES CC1=C(N=CC=C1)N

Specifications

Color White-Yellow
Boiling Point 221°C
Quantity 25 g
UN Number 2811
Formula Weight 108.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Amino-3-methylpyridine
RTECSNumber US1850000
TSCA Yes