missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%

Catalog No. p-7036473
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54773MD 250 mg
AAH5477303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54773MD Supplier Thermo Scientific Chemicals Supplier No. H54773MD

CAS: 1111637-91-2 | C12H19BN2O2 | 234.106 g/mol

It is used as primary and secondary pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Refrigerated (2-4°C). Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1111637-91-2
Molecular Formula C12H19BN2O2
Molecular Weight (g/mol) 234.106
MDL Number MFCD12923388
InChI Key BWEOUBYDXFBFMA-UHFFFAOYSA-N
Synonym 3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 2-amino-3-methylpyridine-5-boronic acid pinacol ester, 3-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 6-amino-5-methylpyridin-3-ylboronic acid pinacol ester, 6-amino-5-methylpyridine-3-boronic acid pinacol ester, 6-amino-5-methylpyridin-3-yl boronic acid pinacol ester, 3-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, 3-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine
PubChem CID 53216772
IUPAC Name 3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C

Specifications

Melting Point 128°C to 130°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 234.1
Percent Purity 96%
Chemical Name or Material 2-Amino-3-methylpyridine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only