missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Amino-3'-methoxyflavone, 99%
A MAP kinase inhibitor | CAS: 167869-21-8 | C16H13NO3 | 267.28 g/mol
$248.90 - $455.45
Chemical Identifiers
| CAS | 167869-21-8 |
|---|---|
| Molecular Formula | C16H13NO3 |
| Molecular Weight (g/mol) | 267.28 |
| MDL Number | MFCD00671789 |
| InChI Key | QFWCYNPOPKQOKV-UHFFFAOYSA-N |
| Synonym | 2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone |
| PubChem CID | 4713 |
| ChEBI | CHEBI:77954 |
| IUPAC Name | 2-(2-amino-3-methoxyphenyl)chromen-4-one |
| SMILES | COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ64485MA
|
Thermo Scientific Chemicals
J64485MA |
10 mg |
Each for $248.90
|
|
|||||
|
AAJ64485MB
|
Thermo Scientific Chemicals
J64485MB |
25 mg |
Each for $455.45
|
|
|||||
Description
A MAP kinase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 167869-21-8 | |
| 267.28 | |
| QFWCYNPOPKQOKV-UHFFFAOYSA-N | |
| 4713 | |
| 2-(2-amino-3-methoxyphenyl)chromen-4-one |
| C16H13NO3 | |
| MFCD00671789 | |
| 2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone | |
| CHEBI:77954 | |
| COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1 |
Specifications
| 167869-21-8 | |
| C16H13NO3 | |
| 10 mg | |
| QFWCYNPOPKQOKV-UHFFFAOYSA-N | |
| 2-(2-amino-3-methoxyphenyl)chromen-4-one | |
| 4713 | |
| 267.28 | |
| Powder |
| Yellow | |
| MFCD00671789 | |
| 2-2-amino-3-methoxyphenyl-4h-chromen-4-one, 2-2-amino-3-methoxyphenyl-4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl chromen-4-one, 2'-amino-3'-methoxyflavone, unii-sje1io5e3i, sje1io5e3i, chembl35482, 2-2-amino-3-methoxyphenyl-chromen-4-one, 4h-1-benzopyran-4-one, 2-2-amino-3-methoxyphenyl, 2′-amino-3′-methoxyflavone | |
| COC1=CC=CC(=C1N)C1=CC(=O)C2=CC=CC=C2O1 | |
| 267.28 | |
| CHEBI:77954 | |
| 99% | |
| 2'-Amino-3'-methoxyflavone |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only