missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-iodopyridine 98.0+%, TCI America™

Catalog No. A25421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25421G 1 g
A25425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25421G Supplier TCI America Supplier No. A25421G
Only null left

Chemical Identifiers

CAS 104830-06-0
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.013
MDL Number MFCD01651885
InChI Key UUDNBWSHTUFGDQ-UHFFFAOYSA-N
Synonym 2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
PubChem CID 2763151
IUPAC Name 3-iodopyridin-2-amine
SMILES C1=CC(=C(N=C1)N)I

Specifications

Melting Point 91°C
Quantity 1 g
Formula Weight 220.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-iodopyridine